Learn More
Sol-M™ Blood Lancet
Standard blood lancet used for capillary blood sampling.
Supplier:  Sol-M™ LAN002
 Afficher plus de versions de ce produit
Numéro de catalogue. 361060271
Il en reste null
Description
Description
  • Used for capillary blood sampling required in daily diabetes management programs
  • Tri-bevel cut cannula supports proper blood sample size
  • Needle is equipped with a protective cap for increased safety and easier placement
Spécifications
Identifiants chimiques
Lancet
Spécifications
28 G
Lancet
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS