Learn More
Sol-M™ Blood Culture Holder
Catalog No. 361060242
Encompass
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
361060242 Non-sterile
1 options
Catalog No. 361060242 Supplier Sol-M™ Supplier No. 110201050002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Blood Culture Holder works with blood collection needle or set, and blood collection bottles as a system in routine venipuncture procedures.
  • Fits all standard evacuated blood culture bottles
  • Threads compatible with standard blood collection needles
Specifications
Disposable Single-use
Latex Free Yes
For Use With (Equipment) Hypodermic Needle, Safety Needle, Blunt Fill Needle
Material Polypropylene
Sterility Non-sterile
Autoclavable Not Autoclavable
Product Type Blood Culture Holder