missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLM Rabbit anti-Mouse, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
The Bloom syndrome gene product is related to the RecQ subset of DExH box-containing DNA helicases and has both DNA-stimulated ATPase and ATP-dependent DNA helicase activities. Mutations causing Bloom syndrome delete or alter helicase motifs and may disable the 3′-5′ helicase activity. The normal protein may act to suppress inappropriate recombination.
Spécifications
Spécifications
| Antigène | BLM |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugué | Unconjugated |
| Formule | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gène | Blm |
| Numéro d’acquisition du gène | O88700 |
| Alias du gène | BLM |
| Symboles de gène | Blm |
| Afficher plus de résultats |