missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps

Numéro de catalogue. 02991340
Change view
Click to view available options
Neck Finish:
22-400
24-400
38-400
70-400
Product Type:
Lined Cap (Smooth)
No. per Case:
144 Cs.
4000 Cs.
576 Cs.
Packaging:
Bagged
Bulk
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Neck Finish Product Type No. per Case Packaging
02991340 24-400 Lined Cap (Smooth) 4000 Cs. Bulk
02993375 70-400 Lined Cap (Smooth) 144 Cs. Bagged
028832F 38-400 Lined Cap (Smooth) 576 Cs. Bagged
361010478 22-400 Lined Cap (Smooth) 576 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 02991340 Fournisseur Qorpak™ Code fournisseur CAP00175
Il en reste null

Used for food, beverage, medical and chemical applications packed at <120°F. Qorpak™ Black Phenolic Pulp/Vinyl Lined Caps are ideal for general use.

  • HDPE coated paper liner
  • Used for mild acids, alkalis, solvents, alcohols, oils and aqueous products
  • Not recommended for active hydrocarbons or bleaches
  • Available in smooth and ribbed style
  • Available as bulk/bagged packaging

Spécifications

Color Black
Material Phenolic Resin
No. per Case 4000 Cs.
Packaging Bulk
Sterility Non-sterile
Liner Material Pulp/Vinyl
Neck Finish 24-400
Disposable Reusable
Product Type Lined Cap (Smooth)
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Afficher plus de résultats Afficher moins de résultats