missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
$1904.83 - $5116.43
Specifications
| Color | Black |
|---|---|
| Autoclavable | Not Autoclavable |
| Clean Claims | DNase, RNase and Pyrogen free |
| For Use With (Application) | Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications |
| Format | Hanging Tip Rack |
| Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Filtered | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Filtered | Price | Quantity | |||
|
14-223-730
|
Axygen™
HT10CBKHTRS5 |
10 μL | Sterile | Non-filtered |
Case of 5760 Each for $3,745.04
|
|
|||
|
14-223-728
|
Axygen™
HT10CBKHTR5 |
10 μL | Non-sterile | Non-filtered |
Case of 5760 Each for $2,734.74
|
|
|||
|
14-223-732
|
Axygen™
HTF10CBKHTRS5 |
10 μL | Sterile | Filtered |
Case of 5760 Each for $5,116.43
|
|
|||
|
14-223-734
|
Axygen™
HT50CBKHTR5 |
50 μL | Non-sterile | Non-filtered |
Case of 5760 Each for $2,609.65
|
|
|||
|
14-223-736
|
Axygen™
HT50CBKHTRS5 |
50 μL | Sterile | Non-filtered |
Case of 5760 Each for $3,256.00
|
|
|||
|
14-223-738
|
Axygen™
HTF50CBKHTRS5 |
50 μL | Sterile | Filtered |
Case of 5760 Each for $4,529.26
|
|
|||
|
14-223-742
|
Axygen™
HT300CBKHTRS5 |
300 μL | Sterile | Non-filtered |
Case of 5760 Each for $3,159.35
|
|
|||
|
14-223-746
|
Axygen™
HT1000CBKHTR5 |
1000 μL | Non-sterile | Non-filtered |
Case of 3840 Each for $1,904.83
|
|
|||
|
14-223-750
|
Axygen™
HTF1000CBKHTRS5 |
1000 μL | Sterile | Filtered |
Case of 3840 Each for $3,343.61
|
|
|||
|
14-223-744
|
Axygen™
HTF300CBKHTRS5 |
300 μL | Sterile | Filtered |
Case of 5760 Each for $4,529.26
|
|
|||
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
- Available in non-sterile, sterile, nonfiltered, and filtered
- Available as Clear or Conductive black polypropylene tips
- Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
- Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
Specifications
| Black | |
| DNase, RNase and Pyrogen free | |
| Hanging Tip Rack |
| Not Autoclavable | |
| Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications | |
| Polypropylene |