Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
Supplier: Axygen™ HT50CBKHTR5
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
- Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
- Available in non-sterile, sterile, nonfiltered, and filtered
- Available as Clear or Conductive black polypropylene tips
- Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
- Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
Specifications
| 50 μL | |
| Non-sterile | |
| Not Autoclavable | |
| Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications | |
| Automation | |
| Hamilton MICROLAB Nimbus |
| Non-filtered | |
| Black | |
| DNase, RNase and Pyrogen free | |
| Hanging Tip Rack | |
| 5 Racks/Pk, 12 Pks/Cs. | |
| Polypropylene |