missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
GREENER_CHOICE

Catalog No. 14223730
Change view
Click to view available options
Volume (Metric):
10 μL
50 μL
300 μL
1000 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Filtered
14223730 10 μL Sterile Non-filtered
14223728 10 μL Non-sterile Non-filtered
14223732 10 μL Sterile Filtered
14223734 50 μL Non-sterile Non-filtered
14223736 50 μL Sterile Non-filtered
14223738 50 μL Sterile Filtered
14223742 300 μL Sterile Non-filtered
14223746 1000 μL Non-sterile Non-filtered
14223750 1000 μL Sterile Filtered
14223744 300 μL Sterile Filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14223730 Supplier Axygen™ Supplier No. HT10CBKHTRS5

Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations

Axygen offers an alternative choice for your Hamilton™ STARline system.

These tips are:

  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Available in non-sterile, sterile, nonfiltered, and filtered
  • Available as Clear or Conductive black polypropylene tips
  • Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Black
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Format Hanging Tip Rack
Tip Style Automation
Packaging 5 Racks/Pk, 12 Pks/Cs.
For Use With (Equipment) Hamilton MICROLAB Nimbus
Material Polypropylene
Show More Show Less