Learn More
Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™, 5 Pack
Compatible with Hamilton Microlab™ Star Line and Nimbus™ automated liquid handling workstations
Supplier:  Axygen™ HT50CBKHTRS5
 View more versions of this product
Catalog No. 14-223-736
Only null left
Explore available promotions
Description
Description
Axygen offers an alternative choice for your Hamilton™ STARline system.
These tips are:
  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • Available in non-sterile, sterile, nonfiltered, and filtered
  • Available as Clear or Conductive black polypropylene tips
  • Available in 96 well hanging tip rack, nested tip rack, and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations
Specifications
Specifications
50 μL
Sterile
Not Autoclavable
Available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Automation
Hamilton MICROLAB Nimbus
Non-filtered
Black
DNase, RNase and Pyrogen free
Hanging Tip Rack
5 Racks/Pk, 12 Pks/Cs.
Polypropylene
Product Suggestions
Product Suggestions