missing translation for 'onlineSavingsMsg'
Learn More

Biuret, 97%

Catalog No. AAL0081236
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0081236 500 g
AAL0081222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0081236 Supplier Thermo Scientific Chemicals Supplier No. L0081236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-19-0 | C2H5N3O2 | 103.081 g/mol

Biuret is used in protein assay. It is also used as Catalytic agent, Petrochemical additive. It is used in organic synthesis. Biuret (Carbamoylurea) was used as a source of dietary nitrogen in purified diets for steers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biuret is used in protein assay. It is also used as Catalytic agent, Petrochemical additive. It is used in organic synthesis. Biuret (Carbamoylurea) was used as a source of dietary nitrogen in purified diets for steers.

Solubility
Soluble in methanol. Partially soluble in cold water, diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases.

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
MDL Number MFCD00007946
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea

Specifications

Melting Point ∽189°C (decomposition)
Density 1.46
Odor Odorless
Linear Formula H2NCONHCONH2
Quantity 500 g
Beilstein 1703510
Merck Index 14,1309
Solubility Information Soluble in methanol. Partially soluble in cold water,diethyl ether.
SMILES C(=O)(N)NC(=O)N
Molecular Weight (g/mol) 103.081
Formula Weight 103.08
Percent Purity 97%
Chemical Name or Material Biuret
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-559-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only