missing translation for 'onlineSavingsMsg'
Learn More

BisphenolA, ≥99%, MilliporeSigma™ Supelco™

$165.76 - $583.72

Chemical Identifiers

MDL Number MFCD00002366
Synonym 2,2-Bis(4-hydroxyphenyl)propane; 4,4 ′-Isopropylidenediphenol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019326
View Documents
Merck Emd Millipore
23965850G
50 g
Each for $165.76
Only null left
 
111018277
View Documents
Merck Emd Millipore
239658250G
250 g
Each for $583.72
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00002366
2,2-Bis(4-hydroxyphenyl)propane; 4,4 ′-Isopropylidenediphenol

Specifications

158°C to 159°C
220°C (4 mmHg)
Poly Bottle
MFCD00002366
2,2-Bis(4-hydroxyphenyl)propane; 4,4 ′-Isopropylidenediphenol
≥99%
80-05-7
227°C - Closed Cup
(CH3)2 C(C6H4 OH)2
50 g
228.29
Videos
Safety and Handling

Safety and Handling

RTECSNumber : SL6300000

SDS
Documents

Documents