missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BisphenolA, ≥99%, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 239658250G
Specifications
| 158°C to 159°C | |
| 220°C (4 mmHg) | |
| Poly Bottle | |
| MFCD00002366 | |
| 2,2-Bis(4-hydroxyphenyl)propane; 4,4 ′-Isopropylidenediphenol | |
| ≥99% |
| 80-05-7 | |
| 227°C - Closed Cup | |
| (CH3)2 C(C6H4 OH)2 | |
| 250 g | |
| 228.29 |
Chemical Identifiers
| MFCD00002366 | |
| 2,2-Bis(4-hydroxyphenyl)propane; 4,4 ′-Isopropylidenediphenol |
Safety and Handling
RTECSNumber : SL6300000