missing translation for 'onlineSavingsMsg'
Learn More

BisphenolA, ≥99%, MilliporeSigma™ Supelco™

Catalog No. 111019326
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019326 50 g
111018277 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019326 Supplier Merck Emd Millipore Supplier No. 23965850G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00002366
Synonym 2,2-Bis(4-hydroxyphenyl)propane; 4,4 ′-Isopropylidenediphenol

Specifications

Melting Point 158°C to 159°C
CAS 80-05-7
Boiling Point 220°C (4 mmHg)
Flash Point 227°C - Closed Cup
Packaging Poly Bottle
Linear Formula (CH3)2 C(C6H4 OH)2
Quantity 50 g
Formula Weight 228.29
Percent Purity ≥99%
RTECSNumber SL6300000