missing translation for 'onlineSavingsMsg'
Learn More

Bismuth titanium oxide, 99.9% (metals basis)

Catalog No. AA4166918
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4166918 50 g
AA4166930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4166918 Supplier Thermo Scientific Chemicals Supplier No. 04166918

CAS: 12233-34-0 | Bi2O11Ti4 | 785.418 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12233-34-0
Molecular Formula Bi2O11Ti4
Molecular Weight (g/mol) 785.418
MDL Number MFCD01310393
InChI Key TZQKEPWGUIJJIY-UHFFFAOYSA-N
Synonym tetraoxo-2,4,6,8,10-pentaoxa-1,11-dibisma-3,5,7,9-tetratitanaundecane-1,11-dione
PubChem CID 56846076
IUPAC Name oxo-oxobismuthanyloxy-[oxo-[oxo-(oxo(oxobismuthanyloxy)titanio)oxytitanio]oxytitanio]oxytitanium
SMILES O=[Ti](O[Ti](=O)O[Ti](=O)O[Bi]=O)O[Ti](=O)O[Bi]=O

Specifications

Physical Form Powder
Quantity 50 g
Formula Weight 785.42
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Bismuth titanium oxide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only