missing translation for 'onlineSavingsMsg'
Learn More

Bismuth carbonate oxide

Catalog No. AAA1248318
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1248318 50 g
AAA1248330 250 g
AAA124830B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1248318 Supplier Thermo Scientific Chemicals Supplier No. A1248318
Only null left

CAS: 5892-10-4 | CBi2O5 | 509.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5892-10-4
Molecular Formula CBi2O5
Molecular Weight (g/mol) 509.97
MDL Number MFCD00010877
InChI Key MGLUJXPJRXTKJM-UHFFFAOYSA-L
Synonym bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide
PubChem CID 16683095
ChEBI CHEBI:31291
IUPAC Name bis(oxobismuthanyl) carbonate
SMILES C(=O)(O[Bi]=O)O[Bi]=O

Specifications

Quantity 50 g
Merck Index 14,1282
Formula Weight 509.97
Density 6.86
Chemical Name or Material Bismuth carbonate oxide
EINECSNumber 227-567-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only