missing translation for 'onlineSavingsMsg'
Learn More

Bismuth antimonide, 99.99%

Catalog No. AAA1701418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1701418 50 g
AAA1701409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1701418 Supplier Thermo Scientific Chemicals Supplier No. A1701418

CAS: 12323-19-2 | Bi2Sb2 | 661.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12323-19-2
Molecular Formula Bi2Sb2
Molecular Weight (g/mol) 661.48
MDL Number MFCD00014196
InChI Key AEMQIQQWIVNHAU-UHFFFAOYSA-N

Specifications

Quantity 50 g
UN Number UN1549
Formula Weight 330.73
Percent Purity 99.99%
Chemical Name or Material Bismuth antimonide
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only