missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis-Tris, ≥99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219454625
Specifications
| Bis-Tris | |
| 6976-37-0 | |
| Crystalline Powder | |
| C8H19NO5 | |
| 2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2' | |
| C(CO)N(CCO)C(CO)(CO)CO | |
| 209.242 | |
| CHEBI:41250 | |
| ≥99% |
| Cell culture reagent | |
| White | |
| 25 g | |
| MFCD00002853 | |
| OWMVSZAMULFTJU-UHFFFAOYSA-N | |
| 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
| 81462 | |
| 209.2 |
Chemical Identifiers
| 6976-37-0 | |
| 209.242 | |
| OWMVSZAMULFTJU-UHFFFAOYSA-N | |
| 81462 | |
| 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
| C8H19NO5 | |
| MFCD00002853 | |
| 2,2-bis(Hydroxymethyl)-2,2',2''-nitrilotriethanol, 2, 2-bis(Hydroxymethyl)-2, 2' | |
| CHEBI:41250 | |
| C(CO)N(CCO)C(CO)(CO)CO |
Safety and Handling
Recommended Storage : Store at Room Temperature (15-30°C).