missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphoranylidene)ammonium chloride, 97% (dry wt.), water <3%

Catalog No. AAL1433914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1433914 25 g
AAL1433906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1433914 Supplier Thermo Scientific Chemicals Supplier No. L1433914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21050-13-5 | C36H30ClNP2 | 574.04 g/mol

Co-catalyst used with a chiral Cobalt-salen complex for the asymmetric addition of carbon dioxide to propylene oxide. Also used in the preparation of 5-Hydroxy-clethodim Sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Co-catalyst used with a chiral Cobalt-salen complex for the asymmetric addition of carbon dioxide to propylene oxide. Also used in the preparation of 5-Hydroxy-clethodim Sulfoxide.

Solubility
Moderately soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 21050-13-5
Molecular Formula C36H30ClNP2
Molecular Weight (g/mol) 574.04
MDL Number MFCD00151523
InChI Key LVRCYPYRKNAAMX-UHFFFAOYSA-M
Synonym 1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium chloride, bis triphenylphosphoranylidene ammonium chloride, bis triphenylphosphoranylidene ammonium, triphenyl p,p,p-triphenylphosphine imidato-n phosphorus 1+ tetracarbonylcobaltate 1-, bis triphenylphosphine iminium chloride∼ppncl, ppn chloride, ppn cl, ksc491c1r, parent, triphenyl p,p,p-triphenylphosphine imidato phosphorus 1+ chloride
PubChem CID 3036656
IUPAC Name triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium;chloride
SMILES [Cl-].C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 274°C to 276°C
Quantity 25 g
Beilstein 4097979
Solubility Information Moderately soluble in water.
Formula Weight 574.05
Percent Purity 97%
Assay (dry wt.), water <3%
Chemical Name or Material Bis(triphenylphosphoranylidene)ammonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-170-6
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only