missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(triphenylphosphine)palladium(II) Diacetate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 749.09 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(triphenylphosphine)palladium(II) Diacetate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |