missing translation for 'onlineSavingsMsg'
Learn More

[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific™

Catalog No. p-2689351
Change view
Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC213670100 10g Glass bottle
AC213670500 50g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC213670100 Supplier Thermo Scientific Chemicals Supplier No. 213670100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

oxidation reagent

Chemical Identifiers

CAS 2712-78-9
Molecular Formula C10H5F6IO4
Molecular Weight (g/mol) 430.03
MDL Number MFCD00009672
InChI Key PEZNEXFPRSOYPL-UHFFFAOYSA-N
Synonym bis trifluoroacetoxy iodo benzene, pifa, unii-659sfv27xs, bis-trifluoroacetoxyiodobenzene, phenylbis trifluoroacetato-o iodine, bis i,i-trifluoroacetoxy iodobenzene, iodine, phenylbis trifluoroacetato-o, bis trifluoracetoxy-iodobenzene, bis trifluoroacetoxy phenyl iodane
PubChem CID 102317
IUPAC Name [phenyl-(2,2,2-trifluoroacetyl)oxy-$l^{3}-iodanyl] 2,2,2-trifluoroacetate
SMILES C1=CC=C(C=C1)I(OC(=O)C(F)(F)F)OC(=O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Linear Formula (CF3CO2)2IC6H5
Quantity 10g
Fieser 09,54
Merck Index 15, 7406
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvent
Formula Weight 430.03
Percent Purity 98%
Chemical Name or Material [Bis(trifluoroacetoxy)iodo]benzene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-308-

RUO – Research Use Only