missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(tri-tert-butylphosphine)palladium(0) 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 511.06 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(tri-tert-butylphosphine)palladium(0) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |