missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-o-tolylphosphine)palladium(II) Dichloride 90.0+%, TCI America™

Catalog No. B20265G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20265G 5 g
B20261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20265G Supplier TCI America Supplier No. B20265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40691-33-6
Molecular Formula C42H42Cl2P2Pd
Molecular Weight (g/mol) 786.06
MDL Number MFCD00274659
InChI Key GMLMZLGOMMIIMA-UHFFFAOYSA-L
Synonym palladium,dichlorobis tris 2-methylphenyl phosphine
PubChem CID 90656943
IUPAC Name dichloropalladiumbis(ylium); bis(tris(2-methylphenyl)phosphane)
SMILES Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 786.07
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tri-o-tolylphosphine)palladium(II) Dichloride