missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-o-tolylphosphine)palladium(0), Pd 14.9%

Catalog No. AA45814MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45814MD 250 mg
AA4581403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA45814MD Supplier Thermo Scientific Chemicals Supplier No. 045814MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Air Sensitive | CAS: 69861-71-8 | C42H42P2Pd | 715.166 g/mol

Bis(tri-o-tolylphosphine)palladium(0) is used as a catalyst for palladium-catalyzed cross-coupling reactions, Kumada coupling and Heck coupling. It is also used in the amination and arylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69861-71-8
Molecular Formula C42H42P2Pd
Molecular Weight (g/mol) 715.166
MDL Number MFCD12911908
InChI Key CUBIJGNGGJBNOC-UHFFFAOYSA-N
Synonym bis tri-o-tolylphosphine palladium 0, pd o-tol 3p 2, bis tris 2-methylphenyl phosphine palladium, bis tris 2-tolyl phosphine palladium, palladium, bis tris 2-methylphenyl phosphine, bis tris 2-methylphenyl phosphane palladium
PubChem CID 10952654
IUPAC Name palladium;tris(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C.[Pd]

Specifications

Melting Point 220°C to 225°C
Color White to Yellow
Assay Percent Range Pd 14.9%
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 715.16
Physical Form Crystalline
Chemical Name or Material Bis(tri-o-tolylphosphine)palladium(0)
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only