missing translation for 'onlineSavingsMsg'
Learn More

Bis(tetramethylcyclopentadienyl)nickel(II), 98+%

Catalog No. AAH2778403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2778403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2778403 Supplier Thermo Scientific Chemicals Supplier No. H2778403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79019-60-6 | C18H26Ni | 301.099 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79019-60-6
Molecular Formula C18H26Ni
Molecular Weight (g/mol) 301.099
MDL Number MFCD01862460
InChI Key PPBIVTWBQQUEKM-UHFFFAOYSA-N
Synonym nickel 2+ bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide, nickel 2+ ion bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide
PubChem CID 73994274
IUPAC Name nickel(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene
SMILES CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Ni+2]

Specifications

Melting Point 107°C to 112°C
Quantity 1 g
Sensitivity Air and moisture sensitive
Formula Weight 301.1
Percent Purity ≥98%
Chemical Name or Material Bis(tetramethylcyclopentadienyl)nickel(II)
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only