missing translation for 'onlineSavingsMsg'
Learn More

Bis(sulfur Dioxide)-1,4-diazabicyclo[2.2.2]octane Adduct 97.0+%, TCI America™

Catalog No. p-7123268
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39601G 1 g
B39605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39601G Supplier TCI America Supplier No. B39601G

Chemical Identifiers

CAS 119752-83-9
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
InChI Key RWISEVUOFYXWFO-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane-1,4-diium-1,4-disulfinate, 1,4-diazoniabicyclo 2.2.2 octane-1,4-disulfinate, 1,4-diazabicyclo 2.2.2 octane-1,4-diium-1,4-disulf, 1,4-diazabicyclo 2.2.2 octane bis sulfur dioxide adduct, 1,4-diazoniabicyclo 2.2.2 octane-1,4-diylbis dioxosulfur vi, 1,4-diazabicyclo 2.2.2 octane bis sulfur dioxide adduct sulfur, elemental analysis
PubChem CID 75176251
IUPAC Name 1,4-diazoniabicyclo[2.2.2]octane-1,4-disulfinate
SMILES C1C[N+]2(CC[N+]1(CC2)S(=O)[O-])S(=O)[O-]

Specifications

Melting Point 180°C
Color White
Quantity 1 g
Formula Weight 240.29
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(sulfur Dioxide)-1,4-diazabicyclo[2.2.2]octane Adduct
TSCA No
Recommended Storage Refrigerator