missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentafluorophenyl) Carbonate 98.0+%, TCI America™

Catalog No. B36045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36045G Supplier TCI America Supplier No. B36045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 59483-84-0
Molecular Formula C13F10O3
Molecular Weight (g/mol) 394.12
MDL Number MFCD00368353
InChI Key IOVVFSGCNWQFQT-UHFFFAOYSA-N
Synonym bis pentafluorophenyl carbonate, bis perfluorophenyl carbonate, dipentafluorophenylcarbonate, pentafluorophenyl carbonate, di-pentafluorophenyl carbonate, bis 2,3,4,5,6-pentafluorophenyl carbonate, carbonic acid bis pentafluorophenyl ester, dpfpc, acmc-209mds
PubChem CID 2734833
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl) carbonate
SMILES FC1=C(F)C(F)=C(OC(=O)OC2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 5 g
Formula Weight 394.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(pentafluorophenyl) Carbonate