missing translation for 'onlineSavingsMsg'
Learn More

Bis(NHS)PEG9, TCI America™

Catalog No. B468825MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B468825MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B468825MG Supplier TCI America Supplier No. B468825MG
Only null left

Chemical Identifiers

CAS 1008402-79-6
Molecular Formula C30H48N2O17
Molecular Weight (g/mol) 708.711
MDL Number MFCD11041131
InChI Key CQWLSZJYTZVHMU-UHFFFAOYSA-N
Synonym Di(N-succinimidyl) 4,7,10,13,16,19,22,25,28-Nonaoxahentriacontanedioate, 4,7,10,13,16,19,22,25,28-Nonaoxahentriacontanedioic Acid Di(N-succinimidyl) Ester
PubChem CID 77078480
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-[2-[2-[2-[2-[2-[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]propanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(=O)ON2C(=O)CCC2=O

Specifications

Color Yellow
Quantity 25 mg
Formula Weight 708.71
Physical Form Liquid
Chemical Name or Material Bis(NHS)PEG9
TSCA No
Recommended Storage Freezer