missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(N,N-dimethylacetamide) Hydrogen Dibromobromate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 92°C |
| Quantity | 25 g |
| Formula Weight | 414.96 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(N,N-dimethylacetamide) Hydrogen Dibromobromate |
Safety and Handling
| TSCA | No |