missing translation for 'onlineSavingsMsg'
Learn More

Bis[eta-(2,5-norbornadiene)]rhodium(I) Tetrafluoroborate, TCI America™

Catalog No. B2091100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2091100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2091100MG Supplier TCI America Supplier No. B2091100MG

Chemical Identifiers

CAS 36620-11-8
Molecular Formula C14H16BF4Rh
Molecular Weight (g/mol) 373.99
MDL Number MFCD00671775
InChI Key HAYDJWBQWOEERB-UHFFFAOYSA-N
Synonym bis norbornadiene rhodium i tetrafluoroborate, rh nbd 2bf4, bis eta-2,5-norbornadiene rhodium i tetrafluoroborate, bis norbornadiene tetrafluoroborato rhodium, bis bicyclo 2.2.1 hepta-2,5-diene rhodium tetrafluoroborate, rh nbd 2 bf4, bis norbomadiene rhodium i tetrafluoroborate, bis norbornadiene rhodium tetrafluoroborate
PubChem CID 10915722
IUPAC Name λ¹-rhodium(1+) bis(bicyclo[2.2.1]hepta-2,5-diene) tetrafluoroboranuide
SMILES [Rh+].F[B-](F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2

Specifications

Melting Point 140°C
Quantity 100 mg
UN Number 1759
Formula Weight 373.99
Physical Form Crystalline Powder
Chemical Name or Material Bis[eta-(2,5-norbornadiene)]rhodium(I) Tetrafluoroborate