missing translation for 'onlineSavingsMsg'
Learn More

Bis(diisopropylamino)chlorophosphine, Thermo Scientific™

Catalog No. AC321560010
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC321560010 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC321560010 Supplier Thermo Scientific Chemicals Supplier No. 321560010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 56183-63-2
Packaging Glass bottle
Molecular Formula C12H28ClN2P
Linear Formula [((CH3)2CH)2N]2PCl
Quantity 1g
MDL Number MFCD00061482
Synonym bis diisopropylamino chlorophosphine, phosphorodiamidous chloride, tetrakis 1-methylethyl, chloro diisopropylamino phosphanyl diisopropylamine, pubchem6478, acmc-1av77, bisdiisopropylaminochlorophosphine, bis diisopropyamino chlorophosphine, bis di-isopropylamino chlorophosphine, bis n,n-diisopropylamino chlorophosphine, chlorobis n,n-diisopropylamino phosphine
Solubility Information Solubility in water: reacts
InChI Key FEHUTHGOLLQBNW-UHFFFAOYSA-N
SMILES CC(C)N(C(C)C)P(Cl)N(C(C)C)C(C)C
IUPAC Name N-[chloro-[di(propan-2-yl)amino]phosphanyl]-N-propan-2-ylpropan-2-amine
Molecular Weight (g/mol) 266.79
PubChem CID 2733425
Formula Weight 266.8
Chemical Name or Material Bis(diisopropylamino)chlorophosphine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only