missing translation for 'onlineSavingsMsg'
Learn More

Bis(dibenzylideneacetone)palladium

Catalog No. AC291970010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291970010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC291970010 Supplier Thermo Scientific Chemicals Supplier No. 291970010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32005-36-0 | C34H28O2Pd | 575.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
Molecular Weight (g/mol) 575.02
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 150.0°C
Color Black to Brown-Red
Packaging Glass bottle
Linear Formula Pd(C6H5CH=CHC(O)CH=CHC6H5)2
Quantity 1 g
Fieser 15,302
Solubility Information Solubility in water: insoluble.
Formula Weight 575.02
Percent Purity 16 to 21% (Pd)
Physical Form Fine Powder
Chemical Name or Material Bis(dibenzylideneacetone)palladium
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only