missing translation for 'onlineSavingsMsg'
Learn More

Bis(dibenzylideneacetone)palladium(0), TCI America™

Catalog No. B13741G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13741G Supplier TCI America Supplier No. B13741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921

Specifications

Melting Point 150°C
Quantity 1 g
SMILES [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
IUPAC Name bis((1E,4E)-1,5-diphenylpenta-1,4-dien-3-one) palladium
Molecular Weight (g/mol) 575.02
Formula Weight 575.02
Physical Form Crystalline Powder
Chemical Name or Material Bis(dibenzylideneacetone)palladium(0)