missing translation for 'onlineSavingsMsg'
Learn More

Bis(di-tert-butyl-phenylphosphine)palladium(0), 98%

Catalog No. AA45853MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45853MD 250 mg
AA4585303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA45853MD Supplier Thermo Scientific Chemicals Supplier No. 045853MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52359-17-8 | C28H46P2Pd | 551.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52359-17-8
Molecular Formula C28H46P2Pd
Molecular Weight (g/mol) 551.04
MDL Number MFCD15071400
InChI Key KJTQUBRAVOMHKY-UHFFFAOYSA-N
Synonym palladium, bis bis 1,1-dimethylethyl phenylphosphine, di-tert-butyl phenyl phosphane-palladium 2/1, bis di-tert-butyl phenyl phosphane palladium
PubChem CID 71365671
IUPAC Name ditert-butyl(phenyl)phosphane;palladium
SMILES [Pd].CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C

Specifications

Color White
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 551.04
Concentration or Composition (by Analyte or Components) Pd 19.3%
Physical Form Crystalline
Chemical Name or Material Bis (di-tert-butyl-phenylphosphine)palladium (0)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only