missing translation for 'onlineSavingsMsg'
Learn More

Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) 98.0+%, TCI America™

Catalog No. B4837500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4837500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4837500MG Supplier TCI America Supplier No. B4837500MG
Only null left

Specifications

CAS 1233717-68-4
Molecular Formula C32H56N2P2Pd
Quantity 500 mg
MDL Number MFCD15071402
Synonym bis di-tert-butyl 4-dimethylaminophenyl phosphine palladium 0, bis 4-n,n-dimethylamino phenyl di-t-butylphosphino palladium 0, bis 4-di-tert-butylphosphanyl-n,n-dimethylaniline palladium
InChI Key SSPOQURGNAWORH-UHFFFAOYSA-N
SMILES CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C.[Pd]
IUPAC Name 4-ditert-butylphosphanyl-N,N-dimethylaniline;palladium
Molecular Weight (g/mol) 637.182
PubChem CID 46900632
Formula Weight 637.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0)
Show More Show Less