missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclohexanone) oxalyldihydrazone 98,0+%, TCI America™

Catalog No. B047625G
Change view
Click to view available options
Quantité:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B047625G 25 g
B04761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B047625G Supplier TCI America Supplier No. B047625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 370-81-0
Formule moléculaire C14H22N4O2
Poids moléculaire (g/mol) 278.356
Numéro MDL MFCD00001659
Clé InChI DSRJIHMZAQEUJV-UHFFFAOYSA-N
Synonyme cuprizone, cuprizane, biscyclohexanone oxaldihydrazone, ethanedioic acid, bis cyclohexylidenehydrazide, oxalic acid bis cyclohexylidenehydrazide, biscyclohexanone oxalyldihydrazone, bis cyclohexanone oxaldihydrazone, cuprizon, unii-5n16u7e0ao, oxalic acid, bis cyclohexylidenehydrazide
PubChem CID 9723
Nom de l’IUPAC N,N'-bis(cyclohexylidenéné) oxamide
SOURIRES C1CCC(=NNC(=O)C(=O)NN=C2CCCCC2)CC1

Specifications

Nom ou substance chimique Bis(cyclohexanone) Oxalyldihydrazone
Point de fusion 208 °C
Couleur Blanc-jaune
Quantité 25 g
Poids de formule 278.36
Pourcentage de pureté ≥98,0% (T)
Forme physique Poudre cristalline
RTECSNumber RO2520000
TSCA Oui