missing translation for 'onlineSavingsMsg'
Learn More

Bis(carboxymethyl) Trithiocarbonate 98.0+%, TCI America™

Catalog No. T20365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20365G 5 g
T203625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20365G Supplier TCI America Supplier No. T20365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6326-83-6
Molecular Formula C5H6O4S3
Molecular Weight (g/mol) 226.28
MDL Number MFCD00004357
InChI Key GQECANUIPBFPLA-UHFFFAOYSA-N
Synonym bis carboxymethyl trithiocarbonate, trithiocarbodiglycolic acid, acetic acid, 2,2'-carbonothioylbis thio bis, thiocarbonyldithioglycolic acid, 3,5-dithia-4-thioxo-1,7-heptanedioic acid, trithiocarbonic acid bis carboxymethyl ester, 2-carboxymethylthio thioxomethylthio acetic acid, carboxymethyl sulfanyl carbothioyl sulfanyl acetic acid
PubChem CID 80618
IUPAC Name 2-({[(carboxymethyl)sulfanyl]methanethioyl}sulfanyl)acetic acid
SMILES OC(=O)CSC(=S)SCC(O)=O

Specifications

Melting Point 173°C
Color Yellow
Quantity 5 g
Formula Weight 226.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(carboxymethyl) Trithiocarbonate