missing translation for 'onlineSavingsMsg'
Learn More

Bis-Acrylamide Solution, 40%, 37:1, Hoefer™

Catalog No. HBGR437500
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HBGR437500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HBGR437500 Supplier Hoefer Inc Supplier No. GR437 500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polyacrylamide gel electrophoresis (PAGE)

Specifications

CAS 110-26-9
Quantity 500 mL
Assay Percent Range 40%
Molecular Formula C7H10N2O2
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILES C=CC(=O)NCNC(=O)C=C
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
Molecular Weight (g/mol) 154.169
PubChem CID 8041
Grade Molecular Biology Ultrapure
Chemical Name or Material Bis-Acrylamide Solution, 37:1
Show More Show Less