missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-nitrophenyl) Sulfide 99.0+%, TCI America™

Catalog No. D08475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08475G 5 g
D084725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08475G Supplier TCI America Supplier No. D08475G
Only null left

Chemical Identifiers

CAS 1223-31-0
Molecular Formula C12H8N2O4S
Molecular Weight (g/mol) 276.266
MDL Number MFCD00039745
InChI Key ZZTJMQPRKBNGNX-UHFFFAOYSA-N
Synonym bis 4-nitrophenyl sulfide, 4-nitrophenyl sulfide, 4,4'-dinitrodiphenyl sulfide, bis p-nitrophenyl sulfide, p,p'-dinitrodiphenyl sulfide, benzene, 1,1'-thiobis 4-nitro, sulfide, bis p-nitrophenyl, bis 4-nitrophenyl sulphide
PubChem CID 14655
IUPAC Name 1-nitro-4-(4-nitrophenyl)sulfanylbenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])SC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 160°C
Quantity 5 g
Formula Weight 276.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-nitrophenyl) Sulfide
EINECSNumber (3)-1111
RTECSNumber WQ2150000
TSCA Yes