missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-nitrophenyl) Disulfide 97.0+%, TCI America™

Catalog No. B050425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B050425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B050425G Supplier TCI America Supplier No. B050425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 100-32-3
Molecular Formula C12H8N2O4S2
Molecular Weight (g/mol) 308.326
MDL Number MFCD00003573
InChI Key KWGZRLZJBLEVFZ-UHFFFAOYSA-N
Synonym 4-nitrophenyl disulfide, bis 4-nitrophenyl disulfide, 4,4'-dinitrodiphenyl disulfide, 1,2-bis 4-nitrophenyl disulfane, p,p'-dinitrodiphenyl disulfide, bis p-nitrophenyl disulfide, disulfide, bis 4-nitrophenyl, p-nitrophenyl disulfide, di-4-nitrophenyl disulfide
PubChem CID 7496
IUPAC Name 1-nitro-4-[(4-nitrophenyl)disulfanyl]benzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])SSC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 183°C
Color Yellow
Quantity 25 g
Formula Weight 308.33
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-nitrophenyl) Disulfide
RTECSNumber JO1550000
TSCA Yes