missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-nitrophenyl) Carbonate 98.0+%, TCI America™

Catalog No. C14815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14815G 5 g
C148125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14815G Supplier TCI America Supplier No. C14815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5070-13-3
Molecular Formula C13H8N2O7
Molecular Weight (g/mol) 304.214
MDL Number MFCD00007322
InChI Key ACBQROXDOHKANW-UHFFFAOYSA-N
Synonym bis 4-nitrophenyl carbonate, bis p-nitrophenyl carbonate, 4,4'-dinitrodiphenyl carbonate, carbonic acid, bis 4-nitrophenyl ester, p,p'-dinitrodiphenylcarbonate, carbonic acid bis 4-nitrophenyl ester, carbonic acid, bis p-nitrophenyl ester, npc bis 4-nitrophenyl carbonate
PubChem CID 78756
IUPAC Name bis(4-nitrophenyl) carbonate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 5 g
Formula Weight 304.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-nitrophenyl) Carbonate