missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-nitrophenyl)amine 98.0+%, TCI America™
$145.40 - $436.19
Chemical Identifiers
| CAS | 1821-27-8 |
|---|---|
| Molecular Formula | C12H9N3O4 |
| Molecular Weight (g/mol) | 259.221 |
| MDL Number | MFCD00437044 |
| InChI Key | MTWHRQTUBOTQTE-UHFFFAOYSA-N |
| PubChem CID | 235381 |
| IUPAC Name | 4-nitro-N-(4-nitrophenyl)aniline |
| SMILES | C1=CC(=CC=C1NC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] |
Chemical Identifiers
| 1821-27-8 | |
| 259.221 | |
| MTWHRQTUBOTQTE-UHFFFAOYSA-N | |
| 4-nitro-N-(4-nitrophenyl)aniline |
| C12H9N3O4 | |
| MFCD00437044 | |
| 235381 | |
| C1=CC(=CC=C1NC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] |
Specifications
| 1821-27-8 | |
| C12H9N3O4 | |
| 1 g | |
| MTWHRQTUBOTQTE-UHFFFAOYSA-N | |
| 4-nitro-N-(4-nitrophenyl)aniline | |
| 235381 | |
| ≥98.0% (HPLC) | |
| Bis(4-nitrophenyl)amine |
| 216°C | |
| MFCD00437044 | |
| 4,4′C-Dinitrodiphenylamine | |
| C1=CC(=CC=C1NC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] | |
| 259.221 | |
| 259.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No