CAS RN 1821-27-8
CAS RN 1821-27-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(4-nitrophenyl)amine 98.0+%, TCI America™
CAS: 1821-27-8 Molecular Formula: C12H9N3O4 Molecular Weight (g/mol): 259.221 MDL Number: MFCD00437044 InChI Key: MTWHRQTUBOTQTE-UHFFFAOYSA-N PubChem CID: 235381 IUPAC Name: 4-nitro-N-(4-nitrophenyl)aniline SMILES: C1=CC(=CC=C1NC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]