missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-nitrophenyl)amine 98.0+%, TCI America™
Supplier: TCI America B29601G
Specifications
Bis(4-nitrophenyl)amine | |
216°C | |
MFCD00437044 | |
4,4′C-Dinitrodiphenylamine | |
C1=CC(=CC=C1NC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] | |
259.221 | |
259.22 | |
Crystalline Powder |
1821-27-8 | |
C12H9N3O4 | |
1 g | |
MTWHRQTUBOTQTE-UHFFFAOYSA-N | |
4-nitro-N-(4-nitrophenyl)aniline | |
235381 | |
≥98.0% (HPLC) |
Chemical Identifiers
1821-27-8 | |
259.221 | |
MTWHRQTUBOTQTE-UHFFFAOYSA-N | |
4-nitro-N-(4-nitrophenyl)aniline |
C12H9N3O4 | |
MFCD00437044 | |
235381 | |
C1=CC(=CC=C1NC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-] |
Safety and Handling
TSCA : No