missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-methoxycarbonylphenyl) Terephthalate 97.0+%, TCI America™

Catalog No. T137025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T137025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T137025G Supplier TCI America Supplier No. T137025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99328-23-1
Molecular Formula C24H18O8
Molecular Weight (g/mol) 434.4
MDL Number MFCD00145360
InChI Key XLVVPIVANBPSAY-UHFFFAOYSA-N
Synonym Terephthalic Acid Bis[4-(methoxycarbonyl)phenyl] Ester, DLC
PubChem CID 13878968
IUPAC Name bis(4-methoxycarbonylphenyl) benzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)C(=O)OC3=CC=C(C=C3)C(=O)OC

Specifications

Color White
Quantity 25 g
Formula Weight 434.40
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-methoxycarbonylphenyl) Terephthalate