missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-hydroxyphényl) disulfure 98,0+%, TCI America™

Catalog No. B38275G
Change view
Click to view available options
Quantité:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B38275G 5 g
B38271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38275G Supplier TCI America Supplier No. B38275G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15015-57-3
Formule moléculaire C12H10O2S2
Poids moléculaire (g/mol) 250.33
Numéro MDL MFCD00020164
Clé InChI XGKGITBBMXTKTE-UHFFFAOYSA-N
PubChem CID 280665
Nom de l’IUPAC 4-[(4-hydroxyphényl)disulfanyl]phénol
SOURIRES C1=CC(=CC=C1O)SSC2=CC=C(C=C2)O

Specifications

Nom ou substance chimique Bis(4-hydroxyphenyl) Disulfide
Point de fusion 152 °C
Couleur Jaune
Quantité 5 g
Synonyme 4,4′-Dihydroxydiphenyl Disulfide, 4,4′-Dithiodiphenol
Poids de formule 250.33
Pourcentage de pureté ≥98,0% (GC)
Forme physique Poudre cristalline