missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-fluorophenyl) sulfone, 98+%

Catalog No. AAL0472314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0472314 25 g
AAL0472306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0472314 Supplier Thermo Scientific Chemicals Supplier No. L0472314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 383-29-9 | C12H8F2O2S | 254.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 383-29-9
Molecular Formula C12H8F2O2S
Molecular Weight (g/mol) 254.25
MDL Number MFCD00000350
InChI Key PLVUIVUKKJTSDM-UHFFFAOYSA-N
Synonym 4-fluorophenyl sulfone, bis p-fluorophenyl sulfone, 4,4'-sulfonylbis fluorobenzene, bis 4-fluorophenyl sulfone, 4,4'-difluorodiphenyl sulfone, 4-fluorophenyl sulphone, 4-fluorophenylsulfone, benzene, 1,1'-sulfonylbis 4-fluoro, 4,4'-difluorodiphenyl sulphone
PubChem CID 67842
IUPAC Name 1-fluoro-4-(4-fluorophenyl)sulfonylbenzene
SMILES FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 97°C to 101°C
Odor Odorless
Quantity 25 g
Beilstein 1970911
Formula Weight 254.25
Percent Purity ≥98%
Chemical Name or Material Bis(4-fluorophenyl) sulfone
EINECSNumber 206-847-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only