missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-bromophenyl)acetylene 98.0+%, TCI America™

Catalog No. B39965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39965G 5 g
B39961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39965G Supplier TCI America Supplier No. B39965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2789-89-1
Molecular Formula C14H8Br2
Molecular Weight (g/mol) 336.03
MDL Number MFCD00185147
InChI Key FJQGIJIHOXZMMJ-UHFFFAOYSA-N
Synonym 1,2-bis 4-bromophenyl ethyne, bis 4-bromophenyl acetylene, 1-bromo-4-2-4-bromophenyl ethynyl benzene, bis-4-bromophenyl acetylene, 1,2-bis 4-bromophenyl acetylene, benzene, 1,1'-1,2-ethynediyl bis 4-bromo, 4,4'-dibromotolan, 4,4'-dibromotolane, bis-p-bromophenyl acetylene, 1 pound not2-bis 4-bromophenyl acetylene
PubChem CID 3464242
IUPAC Name 1-bromo-4-[2-(4-bromophenyl)ethynyl]benzene
SMILES BrC1=CC=C(C=C1)C#CC1=CC=C(Br)C=C1

Specifications

Melting Point 187°C
Color Yellow
Quantity 5 g
Formula Weight 336.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-bromophenyl)acetylene