missing translation for 'onlineSavingsMsg'
Learn More

Bis(3,5-dimethylphenyl)phosphine Oxide 96.0+%, TCI America™

Catalog No. B43905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43905G 5 g
B43901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43905G Supplier TCI America Supplier No. B43905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 187344-92-9
Molecular Formula C16H18OP+
Molecular Weight (g/mol) 257.293
MDL Number MFCD08458884
InChI Key LMXRTXPFJNGAAX-UHFFFAOYSA-N
Synonym bis 3,5-dimethylphenyl phosphine oxide, xyl 2p o h, phosphine oxide,bis 3,5-dimethylphenyl, phosphine oxide, bis 3,5-dimethylphenyl, 1-3,5-dimethylphenyl phosphoryl-3,5-dimethylbenzene, 1-3,5-dimethylphenylphosphoroso-3,5-dimethylbenzene, ksc496c0h, bis 3,5-dimethylphenyl phosphi, 3-4-nitrophenyl-beta-alanine, awaysumoanjulo-uhfffaoysa-n
PubChem CID 12022244
IUPAC Name bis(3,5-dimethylphenyl)-oxophosphanium
SMILES CC1=CC(=CC(=C1)[P+](=O)C2=CC(=CC(=C2)C)C)C

Specifications

Melting Point 110°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 258.30
Percent Purity ≥97.0% (HPLC)
Chemical Name or Material Bis(3,5-dimethylphenyl)phosphine Oxide