missing translation for 'onlineSavingsMsg'
Learn More

Bis[3,5-dibromo-4-(2-hydroxyethoxy)phenyl] Sulfone 96.0+%, TCI America™

Catalog No. B157225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B157225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B157225G Supplier TCI America Supplier No. B157225G
Only null left

Chemical Identifiers

CAS 53714-39-9
Molecular Formula C16H14Br4O6S
Molecular Weight (g/mol) 653.958
MDL Number MFCD00152428
InChI Key OFBQIWFJRDGFEK-UHFFFAOYSA-N
Synonym Bis[4-(2-hydroxyethoxy)-3,5-dibromophenyl] Sulfone
PubChem CID 104566
IUPAC Name 2-[2,6-dibromo-4-[3,5-dibromo-4-(2-hydroxyethoxy)phenyl]sulfonylphenoxy]ethanol
SMILES C1=C(C=C(C(=C1Br)OCCO)Br)S(=O)(=O)C2=CC(=C(C(=C2)Br)OCCO)Br

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 653.96
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis[3,5-dibromo-4-(2-hydroxyethoxy)phenyl] Sulfone
EINECSNumber (3)-3206
TSCA No