missing translation for 'onlineSavingsMsg'
Learn More

Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Oxalate 98.0+%, TCI America™

Catalog No. O02365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02365G 5 g
O023625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02365G Supplier TCI America Supplier No. O02365G

Chemical Identifiers

CAS 30431-54-0
Molecular Formula C26H24Cl6O8
Molecular Weight (g/mol) 677.17
MDL Number MFCD00191674
InChI Key TZZLVFUOAYMTHA-UHFFFAOYSA-N
Synonym Oxalic Acid Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Ester, Bis[2,4,5-trichloro-6-(pentyloxycarbonyl)phenyl] Oxalate, Oxalic Acid Bis[2,4,5-trichloro-6-(pentyloxycarbonyl)phenyl] Ester
PubChem CID 93137
IUPAC Name bis(3,4,6-trichloro-2-pentoxycarbonylphenyl) oxalate
SMILES CCCCCOC(=O)C1=C(C(=CC(=C1Cl)Cl)Cl)OC(=O)C(=O)OC2=C(C(=C(C=C2Cl)Cl)Cl)C(=O)OCCCCC

Specifications

Melting Point 83°C
Color White
Quantity 5 g
Formula Weight 677.17
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Oxalate
EINECSNumber (3)-4136
TSCA Yes