missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) Ether, 99%, Spectrum™ Chemical

Catalog No. 18609082
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609082 4 L Amber Glass Bottle
18609079 250 mL Amber Glass Bottle
18609081 1 L Amber Glass Bottle
18609080 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609082 Supplier Spectrum Chemical Mfg Cor Supplier No. B10994LTPV
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-96-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
InChI Key SBZXBUIDTXKZTM-UHFFFAOYSA-N
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
SMILES COCCOCCOC

Specifications

CAS Max % 1
Boiling Point 162°C (323.6°F)
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material Bis(2-methoxyethyl) Ether

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.